1,3-Diphenyl-1H-pyrazol-5-amine

Supplier: Matrix Scientific

Synonyms: 5-Amino-1,3-diphenylpyrazole

037861-500MG
101844-328EA 193.85 USD
101844-328
1,3-Diphenyl-1H-pyrazol-5-amine
1,3-Diphenyl-1H-pyrazol-5-amine

Hazard: Irritant

Formula: C₁₅H₁₃N₃
MW: 235.29 g/mol
MDL Number: MFCD00084878
CAS Number: 5356-71-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR