3-[(3-Oxo-3,4-dihydro-2H-1,4-benzoxazin-6-yl)sulfonyl]propanoic acid

Supplier: Matrix Scientific

037857-500MG
101844-320EA 484.62 USD
101844-320
3-[(3-Oxo-3,4-dihydro-2H-1,4-benzoxazin-6-yl)sulfonyl]propanoic acid
3-[(3-Oxo-3,4-dihydro-2H-1,4-benzoxazin-6-yl)sulfonyl]propanoic acid

Hazard: Irritant

Formula: C₁₁H₁₁NO₆S
MDL Number: MFCD12027244
CAS Number: 1119451-26-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR