1-[(3-Methyl-2-oxo-2,3-dihydro-1,3-benzoxazol-6-yl)sulfonyl]piperidine-3-carboxylic acid

Supplier: Matrix Scientific

037853-500MG
101844-312EA 290.77 USD
101844-312
1-[(3-Methyl-2-oxo-2,3-dihydro-1,3-benzoxazol-6-yl)sulfonyl]piperidine-3-carboxylic acid
1-[(3-Methyl-2-oxo-2,3-dihydro-1,3-benzoxazol-6-yl)sulfonyl]piperidine-3-carboxylic acid

Hazard: Irritant

Formula: C₁₄H₁₆N₂O₆S
MDL Number: MFCD12027241
CAS Number: 929696-36-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR