[[(3,5-Dimethyl-1H-pyrazol-4-yl)sulfonyl](4-ethoxyphenyl)amino]acetic acid

Supplier: Matrix Scientific

037847-500MG
101844-300EA 290.77 USD
101844-300
[[(3,5-Dimethyl-1H-pyrazol-4-yl)sulfonyl](4-ethoxyphenyl)amino]acetic acid
[[(3,5-Dimethyl-1H-pyrazol-4-yl)sulfonyl](4-ethoxyphenyl)amino]acetic acid

Hazard: Irritant

Formula: C₁₅H₁₉N₃O₅S
MDL Number: MFCD12027237
CAS Number: 1240217-90-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR