[[(1,3-Dimethyl-2,4-dioxo-1,2,3,4-tetrahydro-quinazolin-6-yl)sulfonyl](methyl)amino]acetic acid

Supplier: Matrix Scientific

037845-500MG
101844-296EA 290.77 USD
101844-296
[[(1,3-Dimethyl-2,4-dioxo-1,2,3,4-tetrahydro-quinazolin-6-yl)sulfonyl](methyl)amino]acetic acid
[[(1,3-Dimethyl-2,4-dioxo-1,2,3,4-tetrahydro-quinazolin-6-yl)sulfonyl](methyl)amino]acetic acid

Hazard: Irritant

Formula: C₁₃H₁₅N₃O₆S
MDL Number: MFCD12027235
CAS Number: 773877-46-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR