1-(1H-Imidazol-4-ylsulfonyl)piperidine-4-carboxylic acid

Supplier: Matrix Scientific

037839-500MG
101844-284EA 289.8 USD
101844-284
1-(1H-Imidazol-4-ylsulfonyl)piperidine-4-carboxylic acid
1-(1H-Imidazol-4-ylsulfonyl)piperidine-4-carboxylic acid

Hazard: Irritant

Formula: C₉H₁₃N₃O₄S
MDL Number: MFCD12027230
CAS Number: 899717-99-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR