[(2,1,3-Benzothiadiazol-4-ylsulfonyl)amino](phenyl)acetic acid

Supplier: Matrix Scientific

037820-500MG
101844-246EA 290.77 USD
101844-246
[(2,1,3-Benzothiadiazol-4-ylsulfonyl)amino](phenyl)acetic acid
[(2,1,3-Benzothiadiazol-4-ylsulfonyl)amino](phenyl)acetic acid

Hazard: Irritant

Formula: C₁₄H₁₁N₃O₄S₂
MDL Number: MFCD01080030
CAS Number: 327169-27-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR