3-[(1-Acetyl-2,3-dihydro-1H-indol-5-yl)sulfonyl]propanoic acid

Supplier: Matrix Scientific

037818-500MG
101844-242EA 144.9 USD
101844-242
3-[(1-Acetyl-2,3-dihydro-1H-indol-5-yl)sulfonyl]propanoic acid
3-[(1-Acetyl-2,3-dihydro-1H-indol-5-yl)sulfonyl]propanoic acid

Hazard: Irritant

Formula: C₁₃H₁₅NO₅S
MDL Number: MFCD09893531
CAS Number: 841275-85-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR