3-[(2-Oxo-2,3-dihydro-1,3-benzoxazol-6-yl)sulfonyl]propanoic acid

Supplier: Matrix Scientific

037810-500MG
101844-226EA 290.77 USD
101844-226
3-[(2-Oxo-2,3-dihydro-1,3-benzoxazol-6-yl)sulfonyl]propanoic acid
3-[(2-Oxo-2,3-dihydro-1,3-benzoxazol-6-yl)sulfonyl]propanoic acid

Hazard: Irritant

Formula: C₁₀H₉NO₆S
MDL Number: MFCD03452525
CAS Number: 650603-19-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR