5,7-Dimethyl-6-(3-methylbenzyl)pyrazolo[1,5-a]pyrimidine-3-carboxylic acid

Supplier: Matrix Scientific

037806-500MG
101844-218EA 217.35 USD
101844-218
5,7-Dimethyl-6-(3-methylbenzyl)pyrazolo[1,5-a]pyrimidine-3-carboxylic acid
5,7-Dimethyl-6-(3-methylbenzyl)pyrazolo[1,5-a]pyrimidine-3-carboxylic acid

Hazard: Irritant

Formula: C₁₇H₁₇N₃O₂
MDL Number: MFCD03450667
CAS Number: 656817-41-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR