5,7-Dimethyl-6-(2-methylbenzyl)pyrazolo[1,5-a]pyrimidine-3-carboxylic acid

Supplier: Matrix Scientific

037804-500MG
101844-214EA 217.35 USD
101844-214
5,7-Dimethyl-6-(2-methylbenzyl)pyrazolo[1,5-a]pyrimidine-3-carboxylic acid
5,7-Dimethyl-6-(2-methylbenzyl)pyrazolo[1,5-a]pyrimidine-3-carboxylic acid

Hazard: Irritant

Formula: C₁₇H₁₇N₃O₂
MDL Number: MFCD03019705
CAS Number: 440650-70-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR