N-(2-Bromobenzyl)-N-butyl-N-methylamine

Supplier: Matrix Scientific

037795-500MG
101844-196EA 290.77 USD
101844-196
N-(2-Bromobenzyl)-N-butyl-N-methylamine
N-(2-Bromobenzyl)-N-butyl-N-methylamine

Hazard: Irritant

Formula: C₁₂H₁₈BrN
MDL Number: MFCD12027206
CAS Number: 764667-50-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR