N-Methyl-N-(2-methylcyclohexyl)piperidin-4-amine

Supplier: Matrix Scientific

037793-500MG
101844-192EA 289.8 USD
101844-192
N-Methyl-N-(2-methylcyclohexyl)piperidin-4-amine
N-Methyl-N-(2-methylcyclohexyl)piperidin-4-amine

Hazard: Irritant

Formula: C₁₃H₂₆N₂
MDL Number: MFCD12027204
CAS Number: 1119452-90-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR