6-Ethoxy-2,2-dimethylchroman-4-ol

Supplier: Matrix Scientific

037789-500MG
101844-184EA 290.77 USD
101844-184
6-Ethoxy-2,2-dimethylchroman-4-ol
6-Ethoxy-2,2-dimethylchroman-4-ol

Hazard: Irritant

Formula: C₁₃H₁₈O₃
MDL Number: MFCD09882027
CAS Number: 1017118-08-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR