1-Acetyl-N-(3-morpholin-4-ylpropyl)piperidin-4-amine

Supplier: Matrix Scientific

037787-500MG
101844-180EA 217.35 USD
101844-180
1-Acetyl-N-(3-morpholin-4-ylpropyl)piperidin-4-amine
1-Acetyl-N-(3-morpholin-4-ylpropyl)piperidin-4-amine

Hazard: Irritant

Formula: C₁₄H₂₇N₃O₂
MDL Number: MFCD12027199
CAS Number: 1119450-06-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR