1-Acetyl-N-(3-methylbutyl)piperidin-4-amine

Supplier: Matrix Scientific

037785-500MG
101844-176EA 290.77 USD
101844-176
1-Acetyl-N-(3-methylbutyl)piperidin-4-amine
1-Acetyl-N-(3-methylbutyl)piperidin-4-amine

Hazard: Irritant

Formula: C₁₂H₂₄N₂O
MDL Number: MFCD12027197
CAS Number: 1119452-89-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR