3-[(2-Chloro-4-fluorobenzyl)sulfonyl]propanoic acid

Supplier: Matrix Scientific

037781-500MG
101844-168EA 217.35 USD
101844-168
3-[(2-Chloro-4-fluorobenzyl)sulfonyl]propanoic acid
3-[(2-Chloro-4-fluorobenzyl)sulfonyl]propanoic acid

Hazard: Irritant

Formula: C₁₀H₁₀ClFO₄S
MDL Number: MFCD03830350
CAS Number: 702670-31-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR