N-Butyl-1-ethylpiperidin-4-amine
Supplier: Matrix Scientific
037765-500MG
101844-136EA
290.77
USD
101844-136
N-Butyl-1-ethylpiperidin-4-amine
N-Butyl-1-ethylpiperidin-4-amine
Hazard: Irritant
Formula:
C₁₁H₂₄N₂ |
MDL Number:
MFCD11142441 CAS Number: 1019489-97-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...