N-(2-Bromobenzyl)-N-cyclopentyl-N-methylamine

Supplier: Matrix Scientific

037763-500MG
101844-132EA 193.85 USD
101844-132
N-(2-Bromobenzyl)-N-cyclopentyl-N-methylamine
N-(2-Bromobenzyl)-N-cyclopentyl-N-methylamine

Hazard: Irritant

Formula: C₁₃H₁₈BrN
MDL Number: MFCD12027188
CAS Number: 1119450-04-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR