1-[(5-Methyl-2-phenyl-1,3-oxazol-4-yl)methyl]piperidine-4-carboxylic acid
Supplier: Matrix Scientific
037761-500MG
101844-128EA
387.69
USD
101844-128
1-[(5-Methyl-2-phenyl-1,3-oxazol-4-yl)methyl]piperidine-4-carboxylic acid
1-[(5-Methyl-2-phenyl-1,3-oxazol-4-yl)methyl]piperidine-4-carboxylic acid
Hazard: Irritant
Formula:
C₁₇H₂₀N₂O₃ |
MDL Number:
MFCD12027186 CAS Number: 1119452-87-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...