N-[1-(7-Methoxy-1-benzofuran-2-yl)ethyl]-N-(3-morpholin-4-ylpropyl)amine

Supplier: Matrix Scientific

037753-500MG
101844-112EA 290.77 USD
101844-112
N-[1-(7-Methoxy-1-benzofuran-2-yl)ethyl]-N-(3-morpholin-4-ylpropyl)amine
N-[1-(7-Methoxy-1-benzofuran-2-yl)ethyl]-N-(3-morpholin-4-ylpropyl)amine

Hazard: Irritant

Formula: C₁₈H₂₆N₂O₃
MDL Number: MFCD12027179
CAS Number: 1119451-21-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR