1-(2,3-Dimethylcyclohexyl)-1H-benzimidazole-5-carboxylic acid

Supplier: Matrix Scientific

037751-500MG
101844-108EA 290.77 USD
101844-108
1-(2,3-Dimethylcyclohexyl)-1H-benzimidazole-5-carboxylic acid
1-(2,3-Dimethylcyclohexyl)-1H-benzimidazole-5-carboxylic acid

Hazard: Irritant

Formula: C₁₆H₂₀N₂O₂
MDL Number: MFCD12027178
CAS Number: 953806-84-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR