Methyl (3-nitro-1H-1,2,4-triazol-1-yl)acetate

Supplier: Matrix Scientific

037745-500MG
101844-096EA 289.8 USD
101844-096
Methyl (3-nitro-1H-1,2,4-triazol-1-yl)acetate
Methyl (3-nitro-1H-1,2,4-triazol-1-yl)acetate

Hazard: Irritant

Formula: C₅H₆N₄O₄
MDL Number: MFCD01027023
CAS Number: 70965-23-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR