4,4,4-Trifluoro-1-(4-fluorophenyl)butane-1,3-dione

Supplier: Matrix Scientific

Synonyms: 3-(4-Fluorobenzoyl)-1,1,1-trifluoroacetone

037741-500MG
101844-088EA 217.35 USD
101844-088
4,4,4-Trifluoro-1-(4-fluorophenyl)butane-1,3-dione
4,4,4-Trifluoro-1-(4-fluorophenyl)butane-1,3-dione

Hazard: Irritant

Formula: C₁₀H₆F₄O₂
MW: 234.15 g/mol
MDL Number: MFCD03419766
CAS Number: 582-65-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR