1-(4-Chlorophenyl)-4,4,4-trifluoro-1,3-butanedione

Supplier: Matrix Scientific

Synonyms: 3-(4-Chlorobenzoyl)-1,1,1-trifluoroacetone

037737-500MG
101844-080EA 14.95 USD
101844-080
1-(4-Chlorophenyl)-4,4,4-trifluoro-1,3-butanedione
1-(4-Chlorophenyl)-4,4,4-trifluoro-1,3-butanedione

Hazard: Irritant

Formula: C₁₀H₆ClF₃O₂
MW: 250.61 g/mol
MDL Number: MFCD00511276
CAS Number: 18931-60-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR