5-Phenyl-1,3,4-oxadiazol-2-amine

Supplier: Matrix Scientific

Synonyms: 2-Amino-5-phenyl-1,3,4-oxadiazole

037735-500MG
101844-076EA 144.9 USD
101844-076
5-Phenyl-1,3,4-oxadiazol-2-amine
5-Phenyl-1,3,4-oxadiazol-2-amine

Hazard: Irritant

Formula: C₈H₇N₃O
MW: 161.16 g/mol
MDL Number: MFCD00126383
CAS Number: 1612-76-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR