2-Chloro-N-(2-methoxyethyl)acetamide

Supplier: Matrix Scientific

037729-500MG
101844-060EA 146.15 USD
101844-060
2-Chloro-N-(2-methoxyethyl)acetamide
2-Chloro-N-(2-methoxyethyl)acetamide

Hazard: Irritant

Formula: C₅H₁₀ClNO₂
MDL Number: MFCD00996278
CAS Number: 10263-66-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR