3-(1,3-Dimethyl-1H-pyrazol-4-yl)-5-methylisoxazole-4-carboxylic acid
Supplier: Matrix Scientific
037727-500MG
101844-056EA
387.69
USD
101844-056
3-(1,3-Dimethyl-1H-pyrazol-4-yl)-5-methylisoxazole-4-carboxylic acid
3-(1,3-Dimethyl-1H-pyrazol-4-yl)-5-methylisoxazole-4-carboxylic acid
Hazard: Irritant
Formula:
C₁₀H₁₁N₃O₃ |
MDL Number:
MFCD12027175 CAS Number: 1171972-67-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...