1-Boc-4-piperidineacetic acid

Supplier: Matrix Scientific

Synonyms: 2-(1-(tert-Butoxycarbonyl)piperidin-4-yl)acetic acid[1-(tert-Butoxycarbonyl)piperidin-4-yl]acetic acid

037708-1G
101843-994EA 15.38 USD
101843-994
1-Boc-4-piperidineacetic acid
1-Boc-4-piperidineacetic acid

Hazard: Irritant

Formula: C₁₂H₂₁NO₄
MW: 243.3000 g/mol
MDL Number: MFCD00800239
CAS Number: 157688-46-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR