2-[(1,2-Benzisothiazol-3-ylcarbonyl)amino]benzoic acid

Supplier: Matrix Scientific

037704-500MG
101843-986EA 144.9 USD
101843-986
2-[(1,2-Benzisothiazol-3-ylcarbonyl)amino]benzoic acid
2-[(1,2-Benzisothiazol-3-ylcarbonyl)amino]benzoic acid

Hazard: Irritant

Formula: C₁₅H₁₀N₂O₃S
MDL Number: MFCD12027172
CAS Number: 1031627-74-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR