Ethyl (2-formyl-6-methoxyphenoxy)acetate
Supplier: Matrix Scientific
037702-500MG
101843-982EA
146.15
USD
101843-982
Ethyl (2-formyl-6-methoxyphenoxy)acetate
Ethyl (2-formyl-6-methoxyphenoxy)acetate
Hazard: Irritant
Formula:
C₁₂H₁₄O₅ |
MDL Number:
MFCD03945805 CAS Number: 70076-67-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...