1-Acetyl-N-(4-fluorobenzyl)piperidin-4-amine

Supplier: Matrix Scientific

037694-500MG
101843-966EA 217.35 USD
101843-966
1-Acetyl-N-(4-fluorobenzyl)piperidin-4-amine
1-Acetyl-N-(4-fluorobenzyl)piperidin-4-amine

Hazard: Irritant

Formula: C₁₄H₁₉FN₂O
MDL Number: MFCD01452356
CAS Number: 737745-65-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR