1-Acetyl-N-(1,3-benzodioxol-5-ylmethyl)piperidin-4-amine
Supplier: Matrix Scientific
037692-500MG
101843-962EA
387.69
USD
101843-962
1-Acetyl-N-(1,3-benzodioxol-5-ylmethyl)piperidin-4-amine
1-Acetyl-N-(1,3-benzodioxol-5-ylmethyl)piperidin-4-amine
Hazard: Irritant
Formula:
C₁₅H₂₀N₂O₃ |
MDL Number:
MFCD01652924 CAS Number: 864415-46-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...