N-(1,3-Benzodioxol-5-ylmethyl)-1-benzylpiperidin-4-amine

Supplier: Matrix Scientific

037690-500MG
101843-958EA 290.77 USD
101843-958
N-(1,3-Benzodioxol-5-ylmethyl)-1-benzylpiperidin-4-amine
N-(1,3-Benzodioxol-5-ylmethyl)-1-benzylpiperidin-4-amine

Hazard: Irritant

Formula: C₂₀H₂₄N₂O₂
MDL Number: MFCD01653663
CAS Number: 921102-89-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR