1-Benzyl-N-(4-methoxybenzyl)piperidin-4-amine

Supplier: Matrix Scientific

037686-500MG
101843-950EA 193.85 USD
101843-950
1-Benzyl-N-(4-methoxybenzyl)piperidin-4-amine
1-Benzyl-N-(4-methoxybenzyl)piperidin-4-amine

Hazard: Irritant

Formula: C₂₀H₂₆N₂O
MDL Number: MFCD01653620
CAS Number: 415970-67-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR