1-Ethyl-N-(4-methoxybenzyl)piperidin-4-amine
Supplier: Matrix Scientific
037684-500MG
101843-946EA
193.85
USD
101843-946
1-Ethyl-N-(4-methoxybenzyl)piperidin-4-amine
1-Ethyl-N-(4-methoxybenzyl)piperidin-4-amine
Hazard: Irritant
Formula:
C₁₅H₂₄N₂O |
MDL Number:
MFCD01653633 CAS Number: 415971-71-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...