1-Benzyl-N-(2-furylmethyl)piperidin-4-amine

Supplier: Matrix Scientific

037682-500MG
101843-942EA 144.9 USD
101843-942
1-Benzyl-N-(2-furylmethyl)piperidin-4-amine
1-Benzyl-N-(2-furylmethyl)piperidin-4-amine

Hazard: Irritant

Formula: C₁₇H₂₂N₂O
MDL Number: MFCD01653590
CAS Number: 861408-80-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR