5,7-Dimethyl-1H-indazole

Supplier: Matrix Scientific

037674-500MG
101843-926EA 217.35 USD
101843-926
5,7-Dimethyl-1H-indazole
5,7-Dimethyl-1H-indazole

Hazard: Irritant

Formula: C₉H₁₀N₂
MDL Number: MFCD05663433
CAS Number: 43067-41-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR