3-(1H-Indol-3-yl)-2-(2-methoxyethyl)-1-oxo-1,2-dihydroisoquinoline-4-carboxylic acid

Supplier: Matrix Scientific

037644-500MG
101843-868EA 289.8 USD
101843-868
3-(1H-Indol-3-yl)-2-(2-methoxyethyl)-1-oxo-1,2-dihydroisoquinoline-4-carboxylic acid
3-(1H-Indol-3-yl)-2-(2-methoxyethyl)-1-oxo-1,2-dihydroisoquinoline-4-carboxylic acid

Hazard: Irritant

Formula: C₂₁H₁₈N₂O₄
MDL Number: MFCD12027139
CAS Number: 1119449-96-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR