8-Oxo-5,8,13,13a-tetrahydro-6H-isoquino[3,2-a]isoquinoline-13-carboxylic acid

Supplier: Matrix Scientific

037642-500MG
101843-864EA 387.69 USD
101843-864
8-Oxo-5,8,13,13a-tetrahydro-6H-isoquino[3,2-a]isoquinoline-13-carboxylic acid
8-Oxo-5,8,13,13a-tetrahydro-6H-isoquino[3,2-a]isoquinoline-13-carboxylic acid

Hazard: Irritant

Formula: C₁₈H₁₅NO₃
MW: 293.33 g/mol
MDL Number: MFCD12027137
CAS Number: 1119452-84-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR