[4-[(4-Ethylpiperazin-1-yl)sulfonyl]-1-oxoisoquinolin-2(1H)-yl]acetic acid

Supplier: Matrix Scientific

037637-500MG
101843-856EA 387.69 USD
101843-856
[4-[(4-Ethylpiperazin-1-yl)sulfonyl]-1-oxoisoquinolin-2(1H)-yl]acetic acid
[4-[(4-Ethylpiperazin-1-yl)sulfonyl]-1-oxoisoquinolin-2(1H)-yl]acetic acid

Hazard: Irritant

Formula: C₁₇H₂₁N₃O₅S
MDL Number: MFCD05028863
CAS Number: 1024083-52-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR