Ethyl-2,5-dimethyl-4-oxo-3,4-dihydrothieno[2,3-d]pyrimidine-6-carboxylate

Supplier: Matrix Scientific

037635-500MG
101843-852EA 289.8 USD
101843-852
Ethyl-2,5-dimethyl-4-oxo-3,4-dihydrothieno[2,3-d]pyrimidine-6-carboxylate
Ethyl-2,5-dimethyl-4-oxo-3,4-dihydrothieno[2,3-d]pyrimidine-6-carboxylate

Hazard: Irritant

Formula: C₁₁H₁₂N₂O₃S
MDL Number: MFCD00436801
CAS Number: 23903-53-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR