1-(4-Methylphenyl)cyclohexanecarbonitrile

Supplier: Matrix Scientific

037627-500MG
101843-834EA 87.4 USD
101843-834
1-(4-Methylphenyl)cyclohexanecarbonitrile
1-(4-Methylphenyl)cyclohexanecarbonitrile

Hazard: Irritant

Formula: C₁₄H₁₇N
MDL Number: MFCD00066917
CAS Number: 1206-13-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR