6-Azauracil

Supplier: Matrix Scientific

Synonyms: 1,2,4-Triazine-3,5(2H,4H)-dione

037610-25G
101843-792EA 80.5 USD
101843-792
6-Azauracil
6-Azauracil

Hazard: Irritant

Formula: C₃H₃N₃O₂
MW: 113.08 g/mol
MDL Number: MFCD00006456
CAS Number: 461-89-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR