5-Hydantoin acetic acid

Supplier: Matrix Scientific

037608-500MG
101843-784EA 116.92 USD
101843-784
5-Hydantoin acetic acid
5-Hydantoin acetic acid

Hazard: Irritant

Formula: C₅H₆N₂O₄
MDL Number: MFCD00005267
CAS Number: 5427-26-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR