4-Hydroxy-3-(morpholin-4-ylmethyl)benzoic acid

Supplier: Matrix Scientific

037588-500MG
101843-722EA 87.4 USD
101843-722
4-Hydroxy-3-(morpholin-4-ylmethyl)benzoic acid
4-Hydroxy-3-(morpholin-4-ylmethyl)benzoic acid

Hazard: Irritant

Formula: C₁₂H₁₅NO₄
MW: 237.26 g/mol
MDL Number: MFCD00143178
CAS Number: 143269-99-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR