1-Butyl-1H-benzimidazol-2-amine

Supplier: Matrix Scientific

037580-500MG
101843-706EA 290.77 USD
101843-706
1-Butyl-1H-benzimidazol-2-amine
1-Butyl-1H-benzimidazol-2-amine

Hazard: Irritant

Formula: C₁₁H₁₅N₃
MDL Number: MFCD00159973
CAS Number: 91337-45-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR