2-Formylimidazole ≥95%
Supplier: Matrix Scientific
Synonyms:
1H-Imidazole-2-carbaldehyde
, Imidazole-2-carboxaldehyde
037577-5G
101843-698EA
41.54
USD
101843-698
2-Formylimidazole ≥95%
2-Formylimidazole
Hazard: Irritant/Air Sensitive
Formula:
C₄H₄N₂O MW: 96.09 g/mol Melting Pt: ∼ 205 °C (dec.) |
MDL Number:
MFCD00003544 CAS Number: 10111-08-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...