[(4-Allyl-5-phenyl-4H-1,2,4-triazol-3-yl)thio]acetic acid

Supplier: Matrix Scientific

037573-500MG
101843-688EA 116.92 USD
101843-688
[(4-Allyl-5-phenyl-4H-1,2,4-triazol-3-yl)thio]acetic acid
[(4-Allyl-5-phenyl-4H-1,2,4-triazol-3-yl)thio]acetic acid

Hazard: Irritant

Formula: C₁₃H₁₃N₃O₂S
MDL Number: MFCD00430301
CAS Number: 18204-57-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR