Ethyl-3-(1-adamantyl)-3-oxopropanoate

Supplier: Matrix Scientific

037565-500MG
101843-674EA 87.4 USD
101843-674
Ethyl-3-(1-adamantyl)-3-oxopropanoate
Ethyl-3-(1-adamantyl)-3-oxopropanoate

Hazard: Irritant

Formula: C₁₅H₂₂O₃
MDL Number: MFCD00074741
CAS Number: 19386-06-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR